Skip to Content
Merck
All Photos(1)

Key Documents

730920

Sigma-Aldrich

Palmitoyl-L-carnitine-(N-methyl-d3) hydrochloride

99 atom % D, 98% (CP)

Synonym(s):

Hexadecanoyl-L-carnitine (N-methyl-d3) hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23D3H43ClNO4
Molecular Weight:
439.09
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

98% (CP)

form

powder

mp

166 °C (lit.)

mass shift

M+3

SMILES string

Cl.[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC([O-])=O)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C23H45NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)28-21(19-22(25)26)20-24(2,3)4;/h21H,5-20H2,1-4H3;1H/t21-;/m1./s1/i2D3;

InChI key

GAMKNLFIHBMGQT-HZBHYVOGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi Zhou et al.
Analytica chimica acta, 1109, 44-52 (2020-04-08)
The development of quantitative metabolomics approaches for future standardized and translational applications has become increasingly important. Data-independent targeted quantitative metabolomics (DITQM) is a newly proposed method providing ion pair information on 1324 metabolites. However, the quantification of more than 1000

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service