Skip to Content
Merck
All Photos(2)

Key Documents

721344

Sigma-Aldrich

1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester

97%

Synonym(s):

1,2,3,6-tetrahydro-1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-pyridine, 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-methyl-1,2,3,6-tetrahydropyridine, 4,4,5,5-Tetramethyl-2-[1-methyl-1,2,5,6-tetrahydropyridin-4-yl]-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22BNO2
CAS Number:
Molecular Weight:
223.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

72-76 °C

storage temp.

2-8°C

SMILES string

CN1CCC(=CC1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C12H22BNO2/c1-11(2)12(3,4)16-13(15-11)10-6-8-14(5)9-7-10/h6H,7-9H2,1-5H3

InChI key

SQMVRFXDBRYXFQ-UHFFFAOYSA-N

Application

1-Methyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester can be used as a substrate in the study of para-alkenylation reactions of toluene using thianthrene S-oxide and palladium catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

para-Selective arylation and alkenylation of monosubstituted arenes using thianthrene S-oxide as a transient mediator
Chen X, et al.
Chemical Communications (Cambridge, England), 56, 5058-5061 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service