Skip to Content
Merck
All Photos(1)

Key Documents

717932

Sigma-Aldrich

(±)-Geosmin-(10-methyl-d3)

99 atom % D, 97% (CP)

Synonym(s):

2β,6α-Dimethylbicyclo[4,4.0]decan-β-ol-(10-methyl-d3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D3H19O
Molecular Weight:
185.32
UNSPSC Code:
12352001
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % D

Assay

97% (CP)

form

liquid

technique(s)

bio NMR: suitable
mass spectrometry (MS): suitable

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C12CCCCC1(O)C(C)CCC2

InChI

1S/C12H22O/c1-10-6-5-8-11(2)7-3-4-9-12(10,11)13/h10,13H,3-9H2,1-2H3/i2D3

InChI key

JLPUXFOGCDVKGO-BMSJAHLVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service