Skip to Content
Merck
All Photos(1)

Documents

705942

Sigma-Aldrich

1α,25-Dihydroxyvitamin D3 (6,19,19-d3)

97 atom % D, 96% (CP)

Synonym(s):

1α,25-Dihydroxycholecalciferol (6,19,19-d3), Calcitriol (6,19,19-d3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D3H41O3
CAS Number:
Molecular Weight:
419.65
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

97 atom % D

Quality Level

Assay

96% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1[C@@H](O)C[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1/i2D2,11D

InChI key

GMRQFYUYWCNGIN-POLIPXRRSA-N

General description

1α,25-Dihydroxyvitamin D3 [1,25(OH)2D3 or calcitriol] is a metabolite of vitamin D3. It is known to monitor calcium and phosphate metabolism in humans. It also regulates insulin secretion and induces cellular differentiation. 1α,25-Dihydroxyvitamin D3 (6,19,19-d3) is a deuterated vitamin D3 wherein C-6 and C-19 protons are replaced by deuterium.

Application

1α,25-Dihydroxyvitamin D3 (6,19,19-d3) can be used as an internal analytical standard for the quantification of vitamin D3 concentration in dietary supplements, fortified foods or within a complex biological matrix

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The emerging role of vitamin D3 in women?s health
Khadilkar, SS
Journal of Obstetrics and Gynaecology of India, 63, 147-150 (2013)
Vitamin D3: a helpful immuno-modulator
Di R, et al.
Immunology, 134(2), 123-139 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service