Skip to Content
Merck
All Photos(1)

Documents

680249

Sigma-Aldrich

trans-2-Chloromethylvinylboronic acid pinacol ester

97%

Synonym(s):

(E)-2-(3-Chloro-1-propenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, E-2-Chloromethylvinylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16BClO2
CAS Number:
Molecular Weight:
202.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4603

bp

57-62 °C/0.3-0.4 mmHg

density

1.028 g/mL at 25 °C

SMILES string

CC1(C)OB(OC1(C)C)\C=C\CCl

InChI

1S/C9H16BClO2/c1-8(2)9(3,4)13-10(12-8)6-5-7-11/h5-6H,7H2,1-4H3/b6-5+

InChI key

HLBDNUSUVDDICF-AATRIKPKSA-N

Application

trans-2-Chloromethylvinylboronic acid pinacol ester is a common reactant of Suzuki coupling reaction. It can be used to prepare:
  • Readout reagent that helps in providing qualitative and quantitative readouts in enzyme biomarker detection assays.
  • Boronated enynes, which on Au(I) catalyzed cycloisomerization yield various dienyl boronates.
  • α-Methyl-substituted boronates for the allylation of ketones to yield homoallylic tertiary alcohols.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis, and characterization of small-molecule reagents that cooperatively provide dual readouts for triaging and, when necessary, quantifying point-of-need enzyme assays
Brooks AD, et al.
The Journal of Organic Chemistry, 80(21), 10437-10445 (2015)
Mild and general zinc-alkoxide-catalyzed allylations of ketones with allyl pinacol boronates
Fandrick KR, et al.
Organic Letters, 12(17), 3748-3751 (2010)
Gold-catalyzed cycloisomerization reactions of boronated enynes
Lee JCH and Hall DG
Tetrahedron Letters, 52(2), 321-324 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service