Skip to Content
Merck
All Photos(1)

Key Documents

677922

Sigma-Aldrich

[Bis(trifluoromethanesulfonyl)imidate](triphenylphosphine)gold(I) (2:1) toluene adduct

Synonym(s):

[Bis(trifluoromethanesulfonyl)imidate](PPh3)gold(I) (2:1) toluene adduct

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H15AuF6NO4PS2) 2 · C7H8
Molecular Weight:
1570.93
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

reaction suitability

core: gold
reagent type: catalyst

SMILES string

[Au]N(S(=O)(C(F)(F)F)=O)S(=O)(C(F)(F)F)=O.[Au]N(S(=O)(C(F)(F)F)=O)S(=O)(C(F)(F)F)=O.CC1=CC=CC=C1.P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.P(C5=CC=CC=C5)(C6=CC=CC=C6)C7=CC=CC=C7

InChI

1S/2C18H15P.C7H8.2C2F6NO4S2.2Au/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;2*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;;/h2*1-15H;2-6H,1H3;;;;/q;;;2*-1;2*+1

InChI key

SIVCTOVPPFSWEF-UHFFFAOYSA-N

Application

[Bis(trifluoromethanesulfonyl)imidate](triphenylphosphine)gold(I) (2:1) toluene adduct can be used as a catalyst in the 1,3-isomerisation of propargylic alcohols to enones via Meyer-Schuster rearrangement.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Related Content

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service