Skip to Content
Merck
All Photos(1)

Key Documents

637467

Sigma-Aldrich

D-Valine-d8

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2CDCD(NH2)CO2H
CAS Number:
Molecular Weight:
125.20
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

optical activity

[α]20/D -27.5°, c = 8 in 6 M HCl

technique(s)

protein expression: suitable

mp

>295 °C (subl.) (lit.)

mass shift

M+8

SMILES string

[2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])[C@@]([2H])(N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m1/s1/i1D3,2D3,3D,4D

InChI key

KZSNJWFQEVHDMF-UZTPAVETSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wesley C Kephart et al.
Amino acids, 48(3), 779-789 (2015-11-11)
We examined if supplementing trained cyclists (32 ± 2 year, 77.8 ± 2.6 kg, and 7.4 ± 1.2 year training) with 12 g/day (6 g/day L-Leucine, 2 g/day L-Isoleucine and 4 g/day L-Valine) of either branched-chain amino acids (BCAAs, n
Ellen De Paepe et al.
Analytica chimica acta, 1033, 108-118 (2018-09-03)
In recent years, metabolomics has surfaced as an innovative research strategy in human metabolism, whereby selection of the biological matrix and its inherent metabolome is of crucial importance. However, focusing on a single matrix may imply that relevant molecules of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service