Skip to Content
Merck
All Photos(1)

Key Documents

635871

Sigma-Aldrich

PAMAM-succinamic acid dendrimer, 1,4-diaminobutane core, generation 4 solution

10 wt. % in H2O

Synonym(s):

PAMAM-G4-COOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)4NH2]:(G=4);dendri PAMAM(NHCOCH2CH2COOH)64
Molecular Weight:
20646.87
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

description

succinamic acid surface groups

form

solution

mol wt

generation 4

feature

core type 1,4-diaminobutane core (4-carbon core)

concentration

10 wt. % in H2O

no. Surface Groups

64

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Contains 64 surface succinamic acid groups.
Unlike the cationic amine surface functionalized dendrimers, succinamic acid functional dendrimers offer the researcher anionic surface groups, typical carboxylic acid reactivity and the potential to make carboxylate salts for a variety of reaction capabilities.

Packaging

unit weight includes solvent

Legal Information

Licensed for research use only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service