Skip to Content
Merck
All Photos(1)

Key Documents

617180

Sigma-Aldrich

Bis(2-ethylhexyl)phthalate-3,4,5,6-d4

98 atom % D

Synonym(s):

DEHP-3,4,5,6-d4, Deuterated DEHP, Deuterated bis(2-ethylhexyl)phthalate, Dioctyl phthalate-3,4,5,6-d4, Phthalic acid-3,4,5,6-d4 bis(2-ethylhexyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D4H34O4
CAS Number:
Molecular Weight:
394.58
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

bp

231 °C/5 mmHg (lit.)

mp

-50 °C (lit.)

density

0.996 g/mL at 25 °C

mass shift

M+4

storage temp.

room temp

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OCC(CC)CCCC)c(c1[2H])C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3/i11D,12D,15D,16D

InChI key

BJQHLKABXJIVAM-SAQXESPHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(2-ethylhexyl)phthalate-3,4,5,6-d4 is used to investigate the influence of environmentally relevant dose of DEHP on adipogenesis in the human cell culture model SGBS. It is can be used to study the quantify indoor phthalate ester degradation through both biotic and abiotic mechanisms.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaowei Liu et al.
Environmental science and pollution research international, 22(14), 10690-10698 (2015-03-11)
The first nationwide survey of six phthalates (diethyl phthalate (DEP); dimethyl phthalate (DMP); di-n-butyl phthalate (DBP); butyl benzyl phthalate (BBP); bis(2-ethylhexyl) phthalate (DEHP); din-octyl phthalate (DnOP)) in drinking waters from waterworks was conducted across seven geographical zones in China. Of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service