597651
2-Ethynylaniline
98%
Synonym(s):
(o-Aminophenyl)acetylene, 2-Aminophenylacetylene, 2-Aminophenylethyne, 2-Ethynylbenzenamine, 2-Ethynylphenylamine, o-Ethynylaniline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HC≡CC6H4NH2
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.6200 (lit.)
bp
229-230 °C (lit.)
density
1.030 g/mL at 25 °C (lit.)
SMILES string
Nc1ccccc1C#C
InChI
1S/C8H7N/c1-2-7-5-3-4-6-8(7)9/h1,3-6H,9H2
InChI key
ALQPJHSFIXARGX-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
208.9 °F - closed cup
Flash Point(C)
98.3 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
James M Stubbs et al.
Dalton transactions (Cambridge, England : 2003), 48(22), 7928-7937 (2019-05-16)
A series of [Ru(Cp/Cp*)(PR2NR'2)(MeCN)]PF6 complexes were prepared, in which the steric and electronic properties of the primary coordination sphere were varied (R = Ph, t-Bu, Bn; and Cp vs. Cp*). These complexes were catalytically active in the cyclization of alkyne
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service