Skip to Content
Merck
All Photos(2)

Key Documents

561487

Sigma-Aldrich

1,2-Benzisothiazol-3(2H)-one

97%

Synonym(s):

1,2-Benzisothiazolone, 1,2-Benzoisothiazol-3-one, 1,2-benzothiazoline-3-one, 2,3-Dihydro-1,2-benzothiazol-3-one, 3-Hydroxy-1,2-benzisothiazole, Benzisothiazolin-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NOS
CAS Number:
Molecular Weight:
151.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

154-158 °C (lit.)

SMILES string

Oc1nsc2ccccc12

InChI

1S/C7H5NOS/c9-7-5-3-1-2-4-6(5)10-8-7/h1-4H,(H,8,9)

InChI key

DMSMPAJRVJJAGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Benzisothiazol-3(2H)-one can be synthesized by reacting 2-mercaptobenzoic acid with diphenyl phosphoryl azide.

Application

1,2-Benzisothiazol-3(2H)-one can be used as a precursor in the synthesis of 1,2-benzoisothiazolin-3-one (BIT) derivative which can exhibit enhanced biological activities.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convenient Synthesis of 1, 2-Benzisothiazol-3 (2H)-ones by Cyclization Reaction
of Acyl Azide
Chiyoda T, et al.
Synlett, 2000(10), 1427-1428 (2000)
William L Jorgensen et al.
Bioorganic & medicinal chemistry letters, 21(15), 4545-4549 (2011-07-02)
Substituted N-phenylbenzisothiazolones have been investigated as inhibitors of the tautomerase activity of the proinflammatory cytokine MIF (macrophage migration inhibitory factor). Numerous compounds were found to possess antagonist activity in the low micromolar range with the most potent being the 6-hydroxy
Alan Rolfe et al.
Journal of combinatorial chemistry, 11(4), 732-738 (2009-06-10)
A sequential three-component synthesis of functionalized benzisothiazoline-3-acetic acid 1,1-dioxides utilizing a domino Heck-aza-Michael pathway is reported. This one-pot procedure rapidly assembles functionalized benzylsulfonamides, which undergo a palladium-catalyzed, domino, Heck-aza-Michael transformation in an experimentally straightforward manner. This attractive protocol has been
Kristiina Aalto-Korte et al.
Contact dermatitis, 57(6), 365-370 (2007-11-09)
Benzisothiazolinone is used as a slimicide in the manufacture of disposable powder-free polyvinyl chloride (PVC) gloves. We recently reported 6 patients from dentistry and health care probably sensitized to benzisothiazolinone in PVC gloves. The study aimed to investigate how widely
Deepu Alex et al.
Antimicrobial agents and chemotherapy, 56(9), 4630-4639 (2012-06-13)
We have identified four synthetic compounds (DFD-VI-15, BD-I-186, DFD-V-49, and DFD-V-66) from an amino acid-derived 1,2-benzisothiazolinone (BZT) scaffold that have reasonable MIC(50) values against a panel of fungal pathogens. These compounds have no structural similarity to existing antifungal drugs. Three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service