Skip to Content
Merck
All Photos(1)

Key Documents

547328

Sigma-Aldrich

9-Methyladenine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N5
CAS Number:
Molecular Weight:
149.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

300-305 °C (lit.)

SMILES string

Cn1cnc2c(N)ncnc12

InChI

1S/C6H7N5/c1-11-3-10-4-5(7)8-2-9-6(4)11/h2-3H,1H3,(H2,7,8,9)

InChI key

WRXCXOUDSPTXNX-UHFFFAOYSA-N

Gene Information

General description

9-Methyladenine is a derivative of adenine. It belongs to the monoclinic crystal system and P21/c space group.

Application

9-Methyladenine may be used in the preparation of N6-benzoyl-9-methyladenine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gydo C P van Zundert et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 12(10), 1921-1927 (2011-05-28)
IR spectroscopy is employed to study isolated adenine and its derivative 9-methyladenine in both their neutral and protonated forms. The IR spectra of neutral adenine and 9-methyladenine are measured in a molecular beam expansion via IR-UV ion-dip spectroscopy in the
Sulayman A Oladepo et al.
The journal of physical chemistry. B, 115(19), 6149-6156 (2011-04-23)
The photophysics and photochemistry of nucleobases are the factors governing the photostability of DNA and RNA, since they are the UV chromophores in nucleic acids. Because the formation of photoproducts involves structural changes in the excited electronic state, we study
Chris T Middleton et al.
The journal of physical chemistry. A, 111(42), 10460-10467 (2007-10-04)
Vibrational cooling by 9-methyladenine was studied in a series of solvents by femtosecond transient absorption spectroscopy. Signals at UV and near-UV probe wavelengths were assigned to hot ground state population created by ultrafast internal conversion following electronic excitation by a
Excited state spectroscopy and dynamics of isolated adenine and 9-methyladenine.
Luhrs DC, et al.
Physical Chemistry Chemical Physics, 3(10), 1827-1831 (2001)
Szymon Smolarek et al.
Physical chemistry chemical physics : PCCP, 12(48), 15600-15606 (2010-08-03)
High-resolution absorption spectra of adenine, 9-methyladenine and 2-aminopurine in helium nanodroplets have been recorded. In contrast to molecular beam experiments, large variations in linewidths are observed for adenine and 9-methyladenine. At the same time, the spectrum of 2-aminopurine remains sharp

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service