Skip to Content
Merck
All Photos(2)

Key Documents

537926

Sigma-Aldrich

3-(Diethylenetriamino)propyl-functionalized silica gel

200-400 mesh, extent of labeling: 1.3 mmol/g loading

Synonym(s):

Triamine-3 functionalized silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

extent of labeling

1.3 mmol/g loading

particle size

200-400 mesh

pore size

60 Å pore size

storage temp.

2-8°C

SMILES string

O=[SiH2].CCCNCCNCCN

InChI

1S/C7H19N3.H2OSi/c1-2-4-9-6-7-10-5-3-8;1-2/h9-10H,2-8H2,1H3;2H2

InChI key

VLTNBAWJMOOTTQ-UHFFFAOYSA-N

Related Categories

Application

Metal Scavenger: Pd, Pb, Co, Hg, and Ru

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service