Skip to Content
Merck
All Photos(1)

Key Documents

525081

Sigma-Aldrich

Dimethyl phthalate

≥99%

Synonym(s):

DMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1911460
EC Number:
Council of Europe no.:
34
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99%

form

viscous liquid

autoignition temp.

1033 °F

refractive index

n20/D 1.515 (lit.)

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl phthalate (DMP) is a short-chained ester known as a phthalate acid ester. It is majorly used as a plasticizer for the production of polyvinyl chloride (PVC) and in the coating of cellulose films.

Application

DMP can be used to plasticize cellulose acetate which can be further used in the production of laminated safety glass for flight helmets and goggles. It can be used as a plasticizer that forms complexes with polyvinyl alcohol (PVA) and lithium salts for the development of solid polymer electrolyte (SPE) for solar cell (SC) applications. It may also be used as a mixture of polymeric solvents which can be used in the preparation of polyvinylidene fluoride (PVDF).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-ion conduction of plasticized PVA solid polymer electrolytes complexed with various lithium salts.
Rajendran S, et al.
Solid State Ionics, 167(3-4), 335-339 (2004)
Preparation of PVDF hollow fiber membrane from a ternary polymer/solvent/nonsolvent system via thermally induced phase separation (TIPS) method.
Rajabzadeh S, et al.
Separation and Purification Technology, 63(2), 415-423 (2008)
Raman spectroscopic characterization of laminated glass and transparent sheet plastics to amplify a history of early aviation `glass?.
Madden O, et al.
Journal of Raman Spectroscopy, 45(11-12), 1215-1224 (2014)
Photochemical degradation of dimethyl phthalate by Fenton reagent.
Zhao X, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 161(2-3), 215-220 (2004)
L J Xu et al.
Water research, 47(6), 1996-2004 (2013-02-12)
The merits of the combined process of high-frequency ultrasound (US) and catalyst-free ultraviolet irradiation (UV) have been evaluated in this study by investigating the sonophotolytic degradation of dimethyl phthalate (DMP). A 400 kHz ultrasonic system and a photolytic system at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service