511803
4-Bromobenzyl isocyanate
98%
Synonym(s):
1-Bromo-4-(isocyanatomethyl)benzene
About This Item
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.571 (lit.)
bp
266 °C (lit.)
density
1.514 g/mL at 25 °C (lit.)
functional group
amine
bromo
isocyanate
storage temp.
2-8°C
SMILES string
Brc1ccc(CN=C=O)cc1
InChI
1S/C8H6BrNO/c9-8-3-1-7(2-4-8)5-10-6-11/h1-4H,5H2
InChI key
IXWMDGLNJQNMIO-UHFFFAOYSA-N
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service