Skip to Content
Merck
All Photos(2)

Documents

511471

Sigma-Aldrich

Poly(acrylamide-co-acrylic acid) partial sodium salt

Mw 520,000, Mn 150,000 (Typical), acrylamide ~80 wt. %

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

Mn 150,000 (Typical)
Mw 520,000

composition

acrylamide, ~80 wt. %

impurities

≤7.0 wt. % water

viscosity

150-700 cP(25 °C)(lit.)

solubility

water and some protic solvents: soluble

Mw/Mn

3.5

SMILES string

[Na+].NC(=O)C=C.OC(=O)C=C.[O-]C(=O)C=C

InChI

1S/C3H5NO.2C3H4O2.Na/c3*1-2-3(4)5;/h2H,1H2,(H2,4,5);2*2H,1H2,(H,4,5);/q;;;+1/p-1

InChI key

DTBMFUOWAASNDB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Thickening agent, binder, sizing agent, flocculating agent, crosslinker, filtering aid, lubricant, drag reduction and polymer recovery agent.

Features and Benefits

Changes in pH from 3 to 10 have little effect on viscosity. High tolerance for electrolytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service