Skip to Content
Merck
All Photos(1)

Documents

493023

Sigma-Aldrich

3-Methyl-1-penten-4-yn-3-ol

98%

Synonym(s):

Ethynyl methyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
96.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.446 (lit.)

bp

63-65 °C/100 mmHg (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

CC(O)(C=C)C#C

InChI

1S/C6H8O/c1-4-6(3,7)5-2/h1,5,7H,2H2,3H3

InChI key

VBATUBQIYXCZPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3-Methyl-1-penten-4-yn-3-ol is an acetylenic alcohol. It reacts with ruthenium vinyl carbene to form a ten-membered η2-olefin coordinated ruthenacycle. The efficiency of different palladium catalysts for the hydrogenation of 3-methyl-1-penten-4-yn-3-ol under continuous-flow liquid-phase conditions has been evaluated. It undergoes allylic rearrangement to form cis and trans isomers of 3-methyl-2-penten-4-yn-l-ol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

90.0 °F - closed cup

Flash Point(C)

32.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

From the Lindlar Catalyst to Supported Ligand-Modified Palladium Nanoparticles: Selectivity Patterns and Accessibility Constraints in the Continuous-Flow Three-Phase Hydrogenation of Acetylenic Compounds.
Vile G, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 20(20), 5926-5937 (2014)
Allyl rearrangement of 3-methyl-1-penten-4-yn-3-ol.
Skoda A, et al.
Chemical Papers, 43(6), 743-751 (1989)
1,2-Migration in the reactions of ruthenium vinyl carbene with propargyl alcohols.
Zhou X, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 1(9), 1077-1082 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service