Skip to Content
Merck
All Photos(1)

Key Documents

492248

Sigma-Aldrich

Dimethyl succinate-2,2,3,3-d4

98 atom % D

Synonym(s):

Dimethyl butanedioate-2,2,3,3-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CCD2CD2CO2CH3
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

refractive index

n20/D 1.419 (lit.)

bp

200 °C (lit.)

mp

18-19 °C (lit.)

density

1.148 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]C([2H])(C(=O)OC)C([2H])([2H])C(=O)OC

InChI

1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3/i3D2,4D2

InChI key

MUXOBHXGJLMRAB-KHORGVISSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service