Skip to Content
Merck
All Photos(1)

Documents

488380

Sigma-Aldrich

Sodium butyrate-13C4

99 atom % 13C

Synonym(s):

Butyric acid-13C4 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)213CO2Na
CAS Number:
Molecular Weight:
114.06
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

form

solid

mp

250-253 °C (lit.)

mass shift

M+4

SMILES string

[Na+].[13CH3][13CH2][13CH2][13C]([O-])=O

InChI

1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1/i1+1,2+1,3+1,4+1;

InChI key

MFBOGIVSZKQAPD-UJNKEPEOSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annabell Bachem et al.
Immunity, 51(2), 285-297 (2019-07-06)
Interactions with the microbiota influence many aspects of immunity, including immune cell development, differentiation, and function. Here, we examined the impact of the microbiota on CD8+ T cell memory. Antigen-activated CD8+ T cells transferred into germ-free mice failed to transition into long-lived
Petia Kovatcheva-Datchary et al.
Cell reports, 26(13), 3772-3783 (2019-03-28)
The gut microbiota can modulate human metabolism through interactions with macronutrients. However, microbiota-diet-host interactions are difficult to study because bacteria interact in complex food webs in concert with the host, and many of the bacteria are not yet characterized. To
Dan Vershkov et al.
Cell reports, 26(10), 2531-2539 (2019-03-07)
Fragile X syndrome (FXS) is caused primarily by a CGG repeat expansion in the FMR1 gene that triggers its transcriptional silencing. In order to investigate the regulatory layers involved in FMR1 inactivation, we tested a collection of chromatin modulators for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service