487694
4-Hydroxy-4-methyl-2-pentanone-d12
98 atom % D
Synonym(s):
Diacetone alcohol-d12
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CD3)2C(OD)CD2COCD3
CAS Number:
Molecular Weight:
128.23
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
98 atom % D
Quality Level
Assay
99% (CP)
refractive index
n20/D 1.423 (lit.)
bp
166 °C (lit.)
density
1.026 g/mL at 25 °C
mass shift
M+12
SMILES string
[2H]OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C(=O)C([2H])([2H])[2H]
InChI
1S/C6H12O2/c1-5(7)4-6(2,3)8/h8H,4H2,1-3H3/i1D3,2D3,3D3,4D2,8D
InChI key
SWXVUIWOUIDPGS-YDUIPVTMSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
136.4 °F - closed cup
Flash Point(C)
58.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service