Skip to Content
Merck
All Photos(1)

Key Documents

481653

Sigma-Aldrich

Poly(dimethylsiloxane), chlorine terminated

average Mn ~3,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H13ClO2Si2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

mol wt

average Mn ~3,000

refractive index

n20/D 1.406 (lit.)

viscosity

~3 cSt(lit.)

bp

>177 °C (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)=O.C[Si](C)(O)Cl

InChI key

NBGITOSHNULLLS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service