Skip to Content
Merck
All Photos(1)

Key Documents

47317

Sigma-Aldrich

Fmoc-Lys(Fmoc)-OH

≥98.0% (HPLC)

Synonym(s):

Nα,Nε-di-Fmoc-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H34N2O6
CAS Number:
Molecular Weight:
590.66
Beilstein:
6034060
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](CCCCNC(=O)OCC1c2ccccc2-c3ccccc13)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C36H34N2O6/c39-34(40)33(38-36(42)44-22-32-29-17-7-3-13-25(29)26-14-4-8-18-30(26)32)19-9-10-20-37-35(41)43-21-31-27-15-5-1-11-23(27)24-12-2-6-16-28(24)31/h1-8,11-18,31-33H,9-10,19-22H2,(H,37,41)(H,38,42)(H,39,40)/t33-/m0/s1

InChI key

BMJRTKDVFXYEFS-XIFFEERXSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex Abramov et al.
Molecules (Basel, Switzerland), 24(1) (2018-12-24)
In this work we have investigated the potential benefits of using supramolecular gel networks as reaction media to carry out air-sensitive metal-free light-induced trifluoromethylation of six-membered (hetero)arenes under aerobic conditions. This reaction was performed at room temperature (RT) using sodium
Joseph R Merrill et al.
Biomaterials, 84, 241-249 (2016-02-04)
Tumor angiogenesis, the formation of new tumor blood supply, has been recognized as a hallmark of cancer and represents an important target for clinical management of various angiogenesis-dependent solid tumors. Previously, by screening a bacteriophage peptide library we have discovered
Shafq Al-Azzawi et al.
International journal of molecular sciences, 19(10) (2018-10-21)
Alzheimer's disease (AD) is a progressive brain disorder and age-related disease characterised by abnormal accumulation of β-amyloid (Aβ). The development of drugs to combat AD is hampered by the lack of therapeutically-active molecules able to cross the blood-brain barrier (BBB).
Andrew G Cheetham et al.
Acta pharmacologica Sinica, 38(6), 874-884 (2017-03-07)
The conjugation of small molecular hydrophobic anticancer drugs onto a short peptide with overall hydrophilicity to create self-assembling drug amphiphiles offers a new prodrug strategy, producing well-defined, discrete nanostructures with a high and quantitative drug loading. Here we show the
Aswathi R Hegde et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 102, 237-249 (2017-03-13)
The aim of this study was to evaluate skin delivery of ketoprofen when covalently tethered to mildly cationic (2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service