Skip to Content
Merck
All Photos(1)

Key Documents

447854

Sigma-Aldrich

Diethyl 5-(hydroxymethyl)isophthalate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C6H3-1,3-(CO2C2H5)2
CAS Number:
Molecular Weight:
252.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%

Quality Level

Assay

98%

form

solid

mp

82-85 °C (lit.)

functional group

ester
hydroxyl

SMILES string

CCOC(=O)c1cc(CO)cc(c1)C(=O)OCC

InChI

1S/C13H16O5/c1-3-17-12(15)10-5-9(8-14)6-11(7-10)13(16)18-4-2/h5-7,14H,3-4,8H2,1-2H3

InChI key

AMRPMZYDTZVKTP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrophobic derivatives of 5-(hydroxymethyl) isophthalic acid that selectively induce apoptosis in leukemia cells but not in fibroblasts.
Galkin A, et al.
Drug Development Research, 69(4), 185-195 (2008)
Stefan Raddatz et al.
Nucleic acids research, 30(21), 4793-4802 (2002-11-01)
We report the synthesis of new phosphoramidite building blocks and their use for the modification of oligonucleotides with hydrazides. The reaction of these hydrazide oligonucleotides with active esters and aldehydes is demonstrated for solution conjugation and immobilization. Compared with the
Surface adsorption of polar end-functionalised polystyrenes.
Narrainen AP, et al.
Soft Matter, 2(11), 981-985 (2006)
Gustav Boije af Gennäs et al.
Journal of medicinal chemistry, 52(13), 3969-3981 (2009-05-15)
Protein kinase C (PKC) is a widely studied molecular target for the treatment of cancer and other diseases. We have approached the issue of modifying PKC function by targeting the C1 domain in the regulatory region of the enzyme. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service