Skip to Content
Merck
All Photos(1)

Documents

446130

Sigma-Aldrich

3-[Tris(trimethylsiloxy)silyl]propyl methacrylate

contains MEHQ + HQ as stabilizer, 98%

Synonym(s):

Tris, (3-Methacryloyloxypropyl)tris(trimethylsiloxy)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2(CH2)3Si[OSi(CH3)3]3
CAS Number:
Molecular Weight:
422.81
Beilstein:
1888727
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<1 mmHg ( 20 °C)

Assay

98%

form

liquid

contains

MEHQ + HQ as stabilizer

refractive index

n20/D 1.419 (lit.)

bp

112-115 °C/0.2 mmHg (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C16H38O5Si4/c1-15(2)16(17)18-13-12-14-25(19-22(3,4)5,20-23(6,7)8)21-24(9,10)11/h1,12-14H2,2-11H3

InChI key

BESKSSIEODQWBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

393.8 °F - closed cup

Flash Point(C)

201 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Furqan A Maulvi et al.
Acta biomaterialia, 86, 350-362 (2019-01-10)
Contact lenses are ideally suited for extended drug delivery to the ocular tissues, but incorporation of any particulate system affects the critical properties of the contact lens. Timolol loading by the conventional soaking method does not significantly alter the critical
Hsiu-Wen Chien et al.
Journal of biomaterials science. Polymer edition, 30(12), 1050-1067 (2019-05-21)
The present work proposes to investigate two series of silicone hydrogel materials for their characterization, water content, surface wettability, transmittance, mechanical property, oxygen permeability (Dk), and bacterial attachment as potential contact lens materials and discuss the relationships between water affinity
Furqan A Maulvi et al.
International journal of pharmaceutics, 566, 513-519 (2019-06-09)
The optical and swelling properties of gatifloxacin-loaded contact lens decrease owing to the precipitation of gatifloxacin (on hydration) in the matrix structure of the contact lens. This paper focuses on the use of Pluronic F68 both inside and outside (in
Ankita R Desai et al.
Materials science & engineering. C, Materials for biological applications, 112, 110885-110885 (2020-05-16)
A fixed combination of bimatoprost/timolol eye drop solution is used to manage the elevated intra-ocular pressure in glaucoma patients, including individuals whose condition is poorly controlled by monotherapy. Eye drop solutions are generally given in high dose, due to poor
Nguyen-Phuong-Dung Tran et al.
Polymers, 11(6) (2019-06-05)
In this study, silicone-based hydrogel contact lenses were prepared by the polymerization of 3-(methacryloyloxy)propyltris(trimethylsiloxy)silane (TRIS), N,N-dimethylacrylamide (DMA), 1-vinyl-2-pyrrolidinone (NVP), and 2-hydroxyethylmethacrylate (HEMA). The properties of silicone hydrogel lenses were analyzed based on the methods such as equilibrium water content, oxygen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service