Skip to Content
Merck
All Photos(1)

Documents

424196

Sigma-Aldrich

2-(Trifluoroacetyl)pyrrole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4F3NO
CAS Number:
Molecular Weight:
163.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

48-50 °C (lit.)

SMILES string

FC(F)(F)C(=O)c1ccc[nH]1

InChI

1S/C6H4F3NO/c7-6(8,9)5(11)4-2-1-3-10-4/h1-3,10H

InChI key

UMVVPYXSJKIFST-UHFFFAOYSA-N

General description

2-(Trifluoroacetyl)pyrrole, an α,α,α-trifluoromethyl ketone is a 2-substituted pyrrole derivative. It has been synthesized by reacting pyrrole with trifluoroacetic anhydride in benzene at 0°C. Its enantioselective hydrogenation to form corresponding alcohol using 5wt.% Pt/Al2O3 catalyst chirally modified by new synthetic chiral amines has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemo and enantioselective hydrogenation of fluorinated ketones on platinum modified with (R)-1-(1-naphthyl) ethylamine derivatives.
Diezi S, et al.
J. Mol. Catal. A: Chem., 239(1), 49-56 (2005)
Preparation of Indazoles and Quinazolines by Catalytic Dehydrogenation.
Burnett JrJ and Ainsworth C.
The Journal of Organic Chemistry, 23(9), 1382-1383 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service