Skip to Content
Merck
All Photos(1)

Documents

406619

Sigma-Aldrich

Hexadecafluorodecanedioic acid

Synonym(s):

Hexadecafluorosebacic acid, Perfluorodecanedioic acid, Perfluorosebacic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CF2)8COOH
CAS Number:
Molecular Weight:
490.09
Beilstein:
1898292
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

mp

151-160 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O

InChI

1S/C10H2F16O4/c11-3(12,1(27)28)5(15,16)7(19,20)9(23,24)10(25,26)8(21,22)6(17,18)4(13,14)2(29)30/h(H,27,28)(H,29,30)

InChI key

YPCSMEGZIYWAAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecafluorodecanedioic acid (Hexadecafluorosebacic acid) is a perfluoroalkanedicarboxylic acid, useful in the preparation of synthetic polymers. Negative surface-assisted laser desorption/ionization (SALDI) mass spectra of hexadecafluorosebacic acid has been reported.

Application

Hexadecafluorodecanedioic acid (Perfluorosebacic acid) may be employed as dopant for the synthesis of three-dimensional boxlike microstructures assembled from 1D-nanofibers of polyaniline (PANI). It may be used for the synthesis of polymeric complex {[Mn(O2C(CF2)8CO2)(phen)2]H2O}n.
Reactant involved in:
  • Studies of hydrogen peroxide disproportionation kinetics of manganese phenanthroline perfluorosebacato polymeric complex
  • Synthesis of cobalt phenanthroline perfluorosebacic acid complex
  • Reactions with zinc salt and phenanthroline

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, crystal structure and catalytic activity for H 2 O 2 disproportionation of the manganese (II) coordination polymer {[Mn (O 2 C (CF 2) 8 CO 2)(phen) 2] H 2 O} n (phen= 1, 10-phenanthroline).
Kani I, et al.
Polyhedron, 27(4), 1238-1247 (2008)
Chemical Ionization of Perfluoroalkanedicarboxylic Acid Compounds through Reduction of Copper (II) Sulfide by Laser Irradiation in Surface-Assisted Laser Desorption/Ionization Mass Spectrometry.
KAGAWA S and YOSHIOKA T.
Journal of Mass Spectrometry : Jms, 57(2) (2009)
3D-boxlike polyaniline microstructures with super-hydrophobic and high-crystalline properties.
Zhu Y, et al.
Polymer, 49(16), 3419-3423 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service