Skip to Content
Merck
All Photos(1)

Key Documents

384410

Sigma-Aldrich

Chlorotrimethylsilane solution

1.0 M in THF

Synonym(s):

Trimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCl
CAS Number:
Molecular Weight:
108.64
Beilstein:
1209232
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in THF

density

0.88 g/mL at 25 °C

SMILES string

C[Si](C)(C)Cl

InChI

1S/C3H9ClSi/c1-5(2,3)4/h1-3H3

InChI key

IJOOHPMOJXWVHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorotrimethylsilane (TMSCl) is a silylating agent employed in the silylation of alcohols, amines, carboxylic acids, thiols, and other functional groups. Silyl group serves as a protecting group for these original functional groups.

Application

Chlorotrimethylsilane (TMSCl) can be used as a reagent:
  • To synthesize allenylsilane via carbolithiation of conjugated enynes with aryllithium.
  • In the reductive coupling of aldehydes and ketones to produce corresponding 1,2 diols in the presence of zirconocene dichloride and magnesium.
  • To prepare 4,4′-bis(trimethylsilyl)bicyclohexyl-2,2′-diene from 1,3-cyclohexadiene by reductive disilylation reaction.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zirconocene dichloride-catalyzed pinacol coupling of aromatic aldehydes and ketones
Lakshmi Kantam M, et al.
Synthetic Communications, 36(10), 1437-1445 (2006)
Carbolithiation of conjugated enynes with aryllithiums in microflow system and applications to synthesis of allenylsilanes
Tomida Y, et al.
Organic Letters, 11(16), 3614-3617 (2009)
Ti-catalyzed reactions of 4, 4?-bis (trimethylsilyl) bicyclohexyl-2, 2?-diene with various electrophiles
A Chahinez, et al.
Tetrahedron Letters, 49(31), 4630-4632 (2008)
Malin Persson et al.
Langmuir : the ACS journal of surfaces and colloids, 26(12), 9927-9936 (2010-03-27)
In the in vitro motility assay, actin filaments are propelled by surface-adsorbed myosin motors, or rather, myosin motor fragments such as heavy meromyosin (HMM). Recently, efforts have been made to develop actomyosin powered nanodevices on the basis of this assay
Ainara Sistiaga et al.
PloS one, 9(6), e101045-e101045 (2014-06-26)
Neanderthal dietary reconstructions have, to date, been based on indirect evidence and may underestimate the significance of plants as a food source. While zooarchaeological and stable isotope data have conveyed an image of Neanderthals as largely carnivorous, studies on dental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service