Skip to Content
Merck
All Photos(2)

Key Documents

381357

Sigma-Aldrich

Gallium(III) bromide

99.999%

Synonym(s):

Gallium tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaBr3
CAS Number:
Molecular Weight:
309.44
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
chunks
solubility:
H2O: slightly soluble(lit.)
Pricing and availability is not currently available.

Quality Level

Assay

99.999%

form

chunks

reaction suitability

reagent type: catalyst
core: gallium

impurities

≤15.0 ppm Trace Metal Analysis

bp

279 °C (lit.)

mp

121.5 °C (lit.)

solubility

H2O: slightly soluble(lit.)

density

3.69 g/mL at 25 °C (lit.)

SMILES string

Br[Ga](Br)Br

InChI

1S/3BrH.Ga/h3*1H;/q;;;+3/p-3

InChI key

SRVXDMYFQIODQI-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Gallium(III) bromide may be used to catalyze the coupling of indoles with phenylacetylene to form 1,1-bis(1H-3-indolyl)-1-phenylethanes.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gallium(III) halide-catalyzed coupling of indoles with phenylacetylene: synthesis of bis(indolyl)phenylethanes
Yadav JS, et al.
Tetrahedron Letters, 45(41), 7577-7579 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service