Skip to Content
Merck
All Photos(3)

Key Documents

379859

Sigma-Aldrich

Potassium hexachloroplatinate(IV)

≥99.99% trace metals basis

Synonym(s):

Dipotassium hexachloroplatinate, Dipotassium platinum hexachloride, Platinic potassium chloride, Platinum potassium chloride, Potassium chloroplatinate, Potassium platinum(IV) chloride, Potassium platinum(IV) hexachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl6
CAS Number:
Molecular Weight:
485.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
powder

Quality Level

Assay

≥99.99% trace metals basis

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

mp

250 °C (dec.) (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.2K.Pt/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

DPAIVKJGTXERIM-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium hexachloroplatinate (K2PtCl6) is a source material for the preparation of platinum based compounds. It can be used in the preparation of platinum colloids by its reduction in an aqueous solution.

Application

K2PtCl6 can be used in the preparation of electrodes for potential usage in biosensors. It can be synthesized on nanostructured platinum that can further be used as a cathode catalyst for the oxygen reduction reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xanthine and hypoxanthine sensors based on xanthine oxidase immobilized on a CuPtCl6 chemically modified electrode and liquid chromatography electrochemical detection
Pei J and Li X
Analytica Chimica Acta, 414(1-2), 205-213 (2000)
Characterization and photonic properties for the Pt-fullerene/TiO2 composites derived from titanium (IV) n-butoxide and C60
Oh W and Ko W
Journal of Industrial and Engineering Chemistry, 15(6), 791-797 (2009)
Carbon-supported Pt? Ag nanostructures as cathode catalysts for oxygen reduction reaction
Feng Y, et al.
Physical Chemistry Chemical Physics, 13(9), 3863-3872 (2011)
Shape-selective preparation and properties of oxalate-stabilized Pt colloid
Fu X, et al.
Langmuir, 18(12), 4619-4624 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service