Skip to Content
Merck
All Photos(2)

Key Documents

364266

Sigma-Aldrich

3-Chloropropyl-functionalized silica gel

230-400 mesh, extent of labeling: ~2.5% loading, matrix active group, ~8% functionalized

Synonym(s):

3-Chloropropyl Silica Gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

form

solid

extent of labeling

~2.5% loading

surface area

~550 m2/g

matrix active group

, ~8% functionalized

particle size

230-400 mesh

pore size

60 Å pore size

SMILES string

O[SiH3].CCCCl

Related Categories

Application

Useful as a support for immobilizing substrates on silica gel.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service