Skip to Content
Merck
All Photos(1)

Key Documents

362824

Sigma-Aldrich

Acetyltrimethylsilane

97%

Synonym(s):

1-(Trimethylsilyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCOCH3
CAS Number:
Molecular Weight:
116.23
Beilstein:
2071361
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.411 (lit.)

density

0.811 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)[Si](C)(C)C

InChI

1S/C5H12OSi/c1-5(6)7(2,3)4/h1-4H3

InChI key

REDWSDBFBCDPNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetyltrimethylsilane is an organosilicon compound that is commonly used as a protective agent for alcohols and amines in organic synthesis. It is also used as a reagent in various reactions, which includes trimethylsilylation, condensation, carbon-carbon bond formation, and reductive amination reactions.

Application

Acetyltrimethylsilane can be used as a reagent in the synthesis of dihydrophenanthrenes via a C-C insertion reaction. It is also used as a source of carbanion in the synthesis of unsymmetrical biaryls by carbanion-induced ring transformation of 2H-pyran-2-ones under mild basic conditions.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 26, 6285-6285 (1985)
Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service