Skip to Content
Merck
All Photos(1)

Key Documents

358673

Sigma-Aldrich

Tetrabutylammonium fluoride on silica gel

~1.5 mmol/g (F-)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(F)
Molecular Weight:
261.46
Beilstein:
3570522
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

capacity

~1.5 mmol/g (F-)

SMILES string

[F-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.FH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

FPGGTKZVZWFYPV-UHFFFAOYSA-M

Related Categories

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehrdad Balandeh et al.
Journal of the Electrochemical Society, 164(9), G99-G103 (2017-09-12)
Electrochemical fluorination of methyl(phenylthio)acetate was achieved using tetrabutylammonium fluoride (TBAF). Electrochemical fluorination was performed under potentiostatic anodic oxidation using an undivided cell in acetonitrile containing TBAF and triflic acid. The influence of several parameters including: oxidation potential, time, temperature, sonication
Santosh Kumar Behera et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 14(12), 2225-2237 (2015-10-31)
The mechanism for the dual emission of 2-(4'-N,N-dimethylaminophenyl)imidazo[4,5-c]pyridine (DMAPIP-c) in protic solvents was investigated by synthesizing and studying its analogues. Theoretical calculations were carried out to corroborate the experimental findings. The deprotonation studies suggest that the enhancement in the TICT
Delia López-Velázquez et al.
Carbohydrate polymers, 125, 224-231 (2015-04-11)
We have synthesized and characterized five members of a homologous series of side chain polymers of hydroxypropyl cellulose esters obtained by homogeneous esterification with 6-[4'-(ethoxycarbonyl)biphenyl-4-yloxy]hexanoic acid. Two acylation procedures were studied. One procedure involved the acid chloride derivative and the
Eric P Gillis et al.
Journal of medicinal chemistry, 58(21), 8315-8359 (2015-07-23)
The role of fluorine in drug design and development is expanding rapidly as we learn more about the unique properties associated with this unusual element and how to deploy it with greater sophistication. The judicious introduction of fluorine into a
Soumen Ghosh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 149, 869-874 (2015-05-24)
A series of Schiff bases synthesized by the condensation of benzohydrazide and -NO2 substituted benzaldehyde have been used as selective fluoride ion sensor. Test paper coated with these synthetic Schiff bases (test kits) can detect fluoride ion selectively with a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service