Skip to Content
Merck
All Photos(2)

Documents

341347

Sigma-Aldrich

Bis(diisopropylamino)chlorophosphine

≥95.0%

Synonym(s):

Chlorobis(N,N-diisopropyl)phosphoramidite, Chlorobis(N,N-diisopropylamino)phosphine, Tetraisopropylphosphorodiamidous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)2CH]2N]2PCl
CAS Number:
Molecular Weight:
266.79
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

mp

100-104 °C (lit.)

functional group

phosphine

SMILES string

CC(C)N(C(C)C)P(Cl)N(C(C)C)C(C)C

InChI

1S/C12H28ClN2P/c1-9(2)14(10(3)4)16(13)15(11(5)6)12(7)8/h9-12H,1-8H3

InChI key

FEHUTHGOLLQBNW-UHFFFAOYSA-N

Application

Bis(diisopropylamino)chlorophosphine has been used in the synthesis of tert-butyl tetraisopropylphosphorodiamidite ligand for palladium-catalyzed Buchwald–Hartwig amination reaction.

It is also can be used to synthesize:
  • acyclic N-phosphonio imine catalyst for selective epoxidations
  • boryl-substituted methylenephosphonium derivatives

Other Notes

Contains varying amounts of diisopropylamine hydrochloride

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Borylated methylenephosphonium salts: precursors of elusive boryl (phosphino) carbenes
Lavigne F, et al.
Journal of the American Chemical Society, 132(26), 8864-8865 (2010)
Novel N-phosphonio imine-catalyzed epoxidation reactions
Prieur D, et al.
Organic Letters, 10(11), 2291-2294 (2008)
Palladium?Catalysed Amination of Aryl?and Heteroaryl Halides Using tert?Butyl Tetraisopropylphosphorodiamidite as an Easily Accessible and Air?Stable Ligand
Roiban GD , et al.
European Journal of Organic Chemistry, 2014(10), 2070-2076 (2014)
Shiyue Fang et al.
Current protocols in nucleic acid chemistry, 82(1), e111-e111 (2020-07-07)
This protocol provides details for the preparation of nucleoside phosphoramidites with 1,3-dithian-2-yl-methyl (Dim) and 1,3-dithian-2-yl-methoxycarbonyl (Dmoc) as protecting groups, and a linker with Dmoc as the cleavable function, then using them for solid phase synthesis of sensitive oligodeoxynucleotides (ODNs). Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service