Skip to Content
Merck
All Photos(3)

Key Documents

339768

Sigma-Aldrich

Tetrakis(acetonitrile)palladium(II) tetrafluoroborate

Synonym(s):

NSC 307191, Palladium(II) tetrafluoroborate tetraacetonitrile complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(CH3CN)4(BF4)2
CAS Number:
Molecular Weight:
444.24
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
core: palladium

reagent type: ligand

mp

230 °C (dec.) (lit.)

SMILES string

[Pd++].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F.F[B-](F)(F)F

InChI

1S/4C2H3N.2BF4.Pd/c4*1-2-3;2*2-1(3,4)5;/h4*1H3;;;/q;;;;2*-1;+2

InChI key

YWMRPVUMBTVUEX-UHFFFAOYSA-N

General description

Tetrakis(acetonitrile)palladium(II) tetrafluoroborate is a stronger Lewis acid.[1] It participates in the preparation of 2:1 complex [Pd(1,2-bis(2′-pyridylethynyl)benzene)2](BF4]2, by Sonogashira cross-coupling reaction.[2]

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Reactant involved in:
  • Reactions where it plays a role as the metal source due to weakly coordinated acetonitrile ligands

Precursor for:
  • Synthesis of dendritic SCS-pincer palladium complexes
  • Palladium complexes of click ligands
  • Dipalladium catalysts for use in Heck cross-coupling, Suzuki cross-coupling, and aldehyde olefination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unique 2:1 complex with a trans-chelating bis-pyridine ligand.
Yi-Zhen Hu et al.
Inorganic chemistry, 41(8), 2296-2300 (2002-04-16)
Medez, M.; Mu?oz, M. P.; Echavarren, A. M.
Journal of the American Chemical Society, 122, 11549-11549 (2000)
Takashi Nishikata et al.
Journal of the American Chemical Society, 132(14), 4978-4979 (2010-03-24)
Cationic palladium(II) catalyst realized facile C-H activation of aryl urea with arylboronic acids at room temperature. This reaction is extremely mild to carry out aromatic C-H activations through electrophilic substitution.
Yun Fu Chan et al.
Dalton transactions (Cambridge, England : 2003), (32)(32), 3538-3545 (2007-08-08)
Extended-chain complexes containing multiple transition metal centres linked by conjugated micro-cyanodiazenido(1-) ligands [N=N-C[triple bond, length as m-dash]N]- have been obtained by reaction of trans-[BrW(dppe)2(N2CN)], , [dppe=1,2-bis(diphenylphosphino)ethane] with dirhodium(II) tetra-acetate, bis(benzonitrile)palladium(II) dichloride, and bis(aqua)M(II) bis(hexafluoroacetylacetonate) (M=Mn, Ni, Cu, Zn): stronger Lewis
Maud Reiter et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(27), 7190-7203 (2006-07-05)
Structurally diverse beta-hydroxyenones are shown to undergo nonoxidative 6-endo-trig ring closure to form highly substituted tetrahydropyranones. Amberlyst-15, Al(ClO(4))(3) x 9 H(2)O and [Pd(MeCN)(4)](BF(4))(2) were found to be suitable catalysts for these intramolecular conjugate additions, preventing side reactions, such as dehydration

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the purity of this chemical? And how much percentage of chloride (Cl-) or bromide ion (Br-) is contented in this chemical?

    1 answer
    1. The purity of this item is not tested for this item. The percentage of Chloride and Bromide is not on the COA and not tested for.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service