Skip to Content
Merck
All Photos(1)

Documents

338141

Sigma-Aldrich

Iron

foil, thickness 0.25 mm, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

foil

resistivity

9.71 μΩ-cm

thickness

0.25 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

Related Categories

General description

Iron foils may be used as a substrate for growing graphene by chemical vapor deposition (CVD). Fragmentation of iron foils, using high power laser shock was investigated for engineering and astrophysical applications.

Quantity

  • 6.25CM2 − 25×25mm (approximately 1.2g)
  • 25CM2 − 50×50 mm (approximately 5g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

High-power laser shock-induced dynamic fragmentation of iron foils.
Morard G, et al.
Physical Review. B, Condensed Matter and Materials Physics, 82(17), 174102-174102 (2010)
Synthesis of large-area, few-layer graphene on iron foil by chemical vapor deposition
Xue Y, et al.
Nano Research, 4(12), 1208-1214 (2011)
Tomasz Pytrus et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 34(203), 263-268 (2013-07-31)
The aim of the study was to analyze the occurrence of anemia in children with newly diagnosed Crohn's disease and 3 months after the initiation of treatment depending on location and activity of the disease. 36 children aged from 2
Batool A Haider et al.
BMJ (Clinical research ed.), 346, f3443-f3443 (2013-06-26)
To summarise evidence on the associations of maternal anaemia and prenatal iron use with maternal haematological and adverse pregnancy outcomes; and to evaluate potential exposure-response relations of dose of iron, duration of use, and haemoglobin concentration in prenatal period with
Wanmohd Azhar Wbin-Wan-Ibrahim et al.
Pakistan journal of pharmaceutical sciences, 26(4), 823-826 (2013-07-03)
Heavy metals in cigarette tobacco such as iron may cause a serious damage on human health. Surveys showed that the accumulation of certain toxic heavy metals like cadmium, mercury, iron is very often due to the effect of smoking. This

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service