Skip to Content
Merck
All Photos(1)

Key Documents

301450

Sigma-Aldrich

4-Bromomethyl-6,7-dimethoxycoumarin

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11BrO4
CAS Number:
Molecular Weight:
299.12
Beilstein:
4750487
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

212-216 °C (lit.)

fluorescence

λex 322; λem 395 (Reaction product with acetic acid)
λex 354 nm; λem 435 nm in methanol

functional group

bromo
ester

SMILES string

COc1cc2OC(=O)C=C(CBr)c2cc1OC

InChI

1S/C12H11BrO4/c1-15-10-4-8-7(6-13)3-12(14)17-9(8)5-11(10)16-2/h3-5H,6H2,1-2H3

InChI key

JGODLBJJCNQFII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromomethyl-6,7-dimethoxycoumarin was used as fluorescent labeling reagent for the reversed-phase HPLC separation and detection of carboxylic acids. It was also used as derivatization reagent:
  • for quantitation of clofibric, bezafibric and fenofibric acids in liver by liquid chromatography
  • in determination of the concentration of tripeptide glutathione by matrix-assisted laser desorption ionization time-of-flight mass spectrometry
  • for determination of carboxylic acids in chromatographic detection

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A P Starling et al.
The Biochemical journal, 304 ( Pt 2), 569-575 (1994-12-01)
On reconstitution of the Ca(2+)-ATPase of skeletal muscle sarcoplasmic reticulum into bilayers of dimyristoleoylphosphatidylcholine [di(C14:1)PC] or dinervonylphosphatidylcholine [di(C24:1)PC] the stoichiometry of Ca2+ binding changes from the usual two Ca2+ ions bound per ATPase molecule to one Ca2+ ion bound per
H I Stefanova et al.
Biochemistry, 31(26), 6023-6031 (1992-07-07)
The (Ca(2+)-Mg2+)-ATPase of sarcoplasmic reticulum was labeled with 4-(bromomethyl)-6,7-dimethoxycoumarin. It was shown that a single cysteine residue (Cys-344) was labeled on the ATPase, with a 25% reduction in steady-state ATPase activity and no reduction in the steady-state rate of hydrolysis
K A Dalton et al.
The Biochemical journal, 329 ( Pt 3), 637-646 (1998-03-07)
The sarcoplasmic reticulum of skeletal muscle contains anionic phospholipids as well as the zwitterionic phosphatidylcholine and phosphatidylethanolamine. Here we study the effects of anionic phospholipids on the activity of the Ca2+-ATPase purified from the membrane. Reconstitution of the Ca2+-ATPase into
Minako Karahashi et al.
Biological & pharmaceutical bulletin, 37(1), 105-112 (2014-01-07)
Fibrates are used in biochemical and pharmacological studies as bioactive tools. Nevertheless, most studies have lacked information concerning the concentrations of fibric acids working inside tissues because a simple and sensitive method is not available for their quantitation. This study
4-Bromomethyl-6,7-dimethoxycoumarin as a fluorescence reagent for precolumn derivatization of 5-fluorouracil compounds in high-performance liquid chromatography.
S Yoshida et al.
Journal of chromatography, 430(1), 156-162 (1988-08-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service