Skip to Content
Merck
All Photos(1)

Documents

279439

Sigma-Aldrich

Glycine-2-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N13CH2CO2H
CAS Number:
Molecular Weight:
76.06
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

form

solid

mp

240 °C (dec.) (lit.)

mass shift

M+1

SMILES string

N[13CH2]C(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1+1

InChI key

DHMQDGOQFOQNFH-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuzana Cieslarova et al.
Talanta, 170, 63-68 (2017-05-16)
Amyotrophic lateral sclerosis (ALS) is a neurodegenerative disease that affects both lower and upper motor neurons, leading to muscle atrophy, paralysis, and death caused by respiratory failure or infectious complications. Altered levels of homocysteine, cysteine, methionine, and glutamic acid have
Selçuk Aslan et al.
Metabolic engineering, 57, 96-109 (2019-09-07)
Microbial biosensors are used to detect the presence of compounds provided externally or produced internally. The latter case is commonly constrained by the need to screen a large library of enzyme or pathway variants to identify those that can efficiently
Volker Döring et al.
ACS synthetic biology, 7(9), 2029-2036 (2018-08-15)
Endowing biotechnological platform organisms with new carbon assimilation pathways is a key challenge for industrial biotechnology. Here we report progress toward the construction of formatotrophic Escherichia coli strains. Glycine and serine, universal precursors of one-carbon compounds oxidized during heterotrophic growth
Vidhi Pareek et al.
Science (New York, N.Y.), 368(6488), 283-290 (2020-04-18)
Metabolons, multiprotein complexes consisting of sequential enzymes of a metabolic pathway, are proposed to be biosynthetic "hotspots" within the cell. However, experimental demonstration of their presence and functions has remained challenging. We used metabolomics and in situ three-dimensional submicrometer chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service