Skip to Content
Merck
All Photos(1)

Key Documents

261548

Sigma-Aldrich

Triethylene glycol dimethacrylate

95%, cross-linking reagent polymerization reactions, methacrylate, 80-120 ppm MEHQ as inhibitor

Synonym(s):

TEGDMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)COO(CH2CH2O)3COC(CH3)=CH2
CAS Number:
Molecular Weight:
286.32
Beilstein:
1797351
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%

Assay

95%

form

liquid

contains

80-120 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.461 (lit.)

bp

170-172 °C/5 mmHg (lit.)

density

1.092 g/mL at 25 °C (lit.)

Ω-end

methacrylate

α-end

methacrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C

InChI

1S/C14H22O6/c1-11(2)13(15)19-9-7-17-5-6-18-8-10-20-14(16)12(3)4/h1,3,5-10H2,2,4H3

InChI key

HWSSEYVMGDIFMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylene glycol dimethacrylate (TEGDMA) is a hydrophilic, low viscosity, difunctional methacrylic monomer employed as a crosslinking agent.

Application

TEGDMA is a monomer typically used in dental resin materials that can cause specific stress responses in eukaryotic cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

332.6 °F - closed cup

Flash Point(C)

167 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrzej Dworak et al.
ACS applied materials & interfaces, 5(6), 2197-2207 (2013-03-02)
Well-defined thermosensitive poly[tri(ethylene glycol) monoethyl ether methacrylate] (P(TEGMA-EE)) brushes were synthesized on a solid substrate by the surface-initiated atom transfer radical polymerization of TEGMA-EE. The polymerization reaction was initiated by 2-bromo-2-methylpropionate groups immobilized on the surface of the wafers. The
Eleftherios G Andriotis et al.
Nanomaterials (Basel, Switzerland), 11(1) (2021-01-17)
Mini-emulsion polymerization was applied for the synthesis of cross-linked polymeric nanoparticles comprised of methyl methacrylate (MMA) and Triethylene Glycol Dimethacrylate (TEGDMA) copolymers, used as matrix-carriers for hosting D-limonene. D-limonene was selected as a model essential oil, well known for its
Helmut Schweikl et al.
Biomaterials, 29(10), 1377-1387 (2008-01-01)
Triethylene glycol dimethacrylate (TEGDMA) is a comonomer that is released from dental resin-based materials into hydrophilic solvents. The compound reduces cell vitality, and causes genotoxicity in mammalian cells in vitro. Here, we used gene expression profiling, combined with pathway analysis
Emel Karaman et al.
The journal of adhesive dentistry, 14(5), 485-492 (2012-06-23)
To evaluate the 24-month clinical performance of cervical restorations using a nanohybrid and a flowable resin composite with a one-step self-etching adhesive. Twenty-one patients with at least one pair of non-carious cervical lesions participated in this study. A total of
Geum-Jun Han et al.
The journal of adhesive dentistry, 14(5), 461-469 (2012-06-23)
To evaluate the effect of applied power on dental ceramic bonding of composite resin using nonthermal atmospheric pressure plasma (APP). A pencil-type APP torch was used to modify the surface chemical composition and hydrophilicity of dental ceramic and to improve

Articles

With dentists placing nearly 100 million dental fillings into patients′ teeth annually in the U.S. alone, polymeric composite restoratives account for a very large share of the biomaterials market.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service