Skip to Content
Merck
All Photos(2)

Key Documents

239437

Sigma-Aldrich

3-Ethoxypropylamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O(CH2)3NH2
CAS Number:
Molecular Weight:
103.16
Beilstein:
1732287
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%

Quality Level

Assay

99%

refractive index

n20/D 1.418 (lit.)

bp

136-138 °C (lit.)

density

0.861 g/mL at 25 °C (lit.)

functional group

amine
ether

SMILES string

CCOCCCN

InChI

1S/C5H13NO/c1-2-7-5-3-4-6/h2-6H2,1H3

InChI key

SOYBEXQHNURCGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Ethoxypropylamine was used to partially open the rigid oxetane chain, during the synthesis of oxetane-based oligomers, to generate some specific properties.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

91.4 °F - DIN 51755 Part 1

Flash Point(C)

33 °C - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and modification of oxetane based oligomers with 3-ethoxypropylamine by focused microwave irradiation.
Baudel V, et al.
Eur. Polymer J., 38(3), 615-618 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service