Skip to Content
Merck
All Photos(5)

Key Documents

226491

Sigma-Aldrich

1,1,3,3-Tetramethylbutyl isocyanide

greener alternative

technical grade

Synonym(s):

tert-Octyl isocyanide, Walborsky’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2C(CH3)2NC
CAS Number:
Molecular Weight:
139.24
Beilstein:
2076559
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

greener alternative product score

old score: 5
new score: 3
Find out more about DOZN™ Scoring

greener alternative product characteristics

Safer Solvents and Auxiliaries
Design for Energy Efficiency
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.422 (lit.)

bp

55-57 °C/11 mmHg (lit.)

density

0.794 g/mL at 25 °C (lit.)

functional group

amine

greener alternative category

storage temp.

2-8°C

SMILES string

CC(C)(C)CC(C)(C)[N+]#[C-]

InChI

1S/C9H17N/c1-8(2,3)7-9(4,5)10-6/h7H2,1-5H3

InChI key

YVPXQMYCTGCWBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Safer Solvents and Auxiliaries”, “Enhanced Energy Efficiency” and “Design for Degradation”. Click here to view its DOZN scorecard.

Application

1,1,3,3-Tetramethylbutyl isocyanide was used in the synthesis of substituted furans and imides.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramazani A and Holagh MV.
Phosphorus, Sulfur, and Silicon and the Related Elements, 184(1), 171-178 (2008)
Esben Thyssen Vestergaard et al.
The Journal of clinical endocrinology and metabolism, 104(7), 2581-2592 (2019-02-07)
Glucagon-like peptide-1 (GLP-1) is an incretin hormone used therapeutically in type 2 diabetes and obesity. The interplay between ambient free fatty acids (FFAs) and GLP-1 remains unclear. Acipimox suppresses adipose tissue lipolysis via activation of the PUMA-G (also known as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service