Skip to Content
Merck
All Photos(3)

Documents

215392

Sigma-Aldrich

Copper(II) perchlorate hexahydrate

98%

Synonym(s):

Cupric perchlorate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(ClO4)2 · 6H2O
CAS Number:
Molecular Weight:
370.54
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

crystalline

reaction suitability

reagent type: oxidant

density

2.225 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O.O.O.O.O.O.[Cu++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Cu.6H2O/c2*2-1(3,4)5;;;;;;;/h2*(H,2,3,4,5);;6*1H2/q;;+2;;;;;;/p-2

InChI key

NHELIHXBJRANPL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The monoclinic structure of copper perchlorate hexahydrate was analyzed by X-ray diffraction. The product may be used as a catalyst during the reaction of 5-methylisophthalaldehyde with 1,3-diaminopropane. Benzaldehyde semicarbazones, reacted with copper perchlorate hexahydrate in acetonitrile, may yield corresponding 1,2,4-triazolin-5-ones.

Application

  • Methanol[1-(methoxymethanimidoyl)-2-(pyridin-2-ylmethyl)guanidine]bis(perchlorato)copper(II).: This study reports the synthesis and crystal structure of a new copper(II) complex with potential applications in materials science and catalysis. The research focuses on the structural characterization of the complex, which is crucial for understanding its reactivity and potential uses in various chemical processes (Meenongwa et al., 2012).
  • Oxidative cleavage of DNA by a dipyridoquinoxaline copper(II) complex in the presence of ascorbic acid.: This research explores the catalytic activity of a copper(II) complex in oxidative DNA cleavage. The findings demonstrate the complex′s potential application in biochemical studies and the development of new catalytic processes for DNA manipulation and analysis (Santra et al., 2002).

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabri Ahmed Cherrak et al.
PloS one, 11(10), e0165575-e0165575 (2016-10-28)
Natural flavonoids such as quercetin, (+)catechin and rutin as well as four methoxylated derivatives of quercetin used as models were investigated to elucidate their impact on the oxidant and antioxidant status of human red blood cells (RBCs). The impact of
The crystal structure of copper perchlorate hexahydrate, Cu (ClO4) 2? 6 H20.
Mani NV and Ramaseshan S
Zeitschrift fur Kristallographie, 115(1-6), 97-109 (1961)
Sze-Wing Ng et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-01)
A new tetradentate mixed aza-thioether macrocyclic ligand 2,6-dithia[7](2,9)-1,10-phenanthrolinophane ([13]ane(phenN2)S2) was successfully synthesized. Reacting metal precursors [Fe(CH3CN)2(OTf)2], Ni(ClO4)2·6H2O, and Cu(ClO4)2·6H2O with one equivalent of [13]ane(phenN2)S2 afforded [Fe([13]ane(phenN2)S2)(OTf)2] (1), [Ni([13]ane(phenN2)S2)](ClO4)2 (2(ClO4)2), and [Cu([13]ane(phenN2)S2)(OH2)](ClO4)2 (3(ClO4)2), respectively. The structures of [13]ane(phenN2)S2 and all of
Complexes of binucleating ligands. III. Novel complexes of a macrocyclic binucleating ligand.
Pilkington NH and Robson R
Australian Journal of Chemistry, 23(11), 2225-2236 (1970)
A study of the mechanism of the oxidative cyclization of benzaldehyde semicarbazones induced by cupric perchlorate in acetonitrile
Noto R, et al.
Journal of Heterocyclic Chemistry, 32(4), 1277-1282 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service