Skip to Content
Merck
All Photos(1)

Key Documents

210137

Sigma-Aldrich

2-(Methylamino)pyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.578 (lit.)

bp

200-201 °C (lit.)

mp

15 °C (lit.)

density

1.052 g/mL at 25 °C (lit.)

SMILES string

CNc1ccccn1

InChI

1S/C6H8N2/c1-7-6-4-2-3-5-8-6/h2-5H,1H3,(H,7,8)

InChI key

SVEUVITYHIHZQE-UHFFFAOYSA-N

General description

Photoreaction of 2-(methylamino)pyridine in low-temperature argon matrix has been investigated by infrared spectroscopy and density functional theory calculation. 2-(Methylamino)pyridine forms copper(II) complexes containing various anions (Cl-, Br-, NO3-, SCN-, ClO4-, or SO42-).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemistry of 2-(methylamino) pyridine in a low-temperature argon matrix: Amino-imino tautomerism and rotational isomerism.
Akai N, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 187(1), 113-118 (2007)
Synthesis, characterization and demonstration of superoxide dismutase-like activity of copper (II) chloride, bromide, nitrate, thiocyanate, sulphate, and perchlorate complexes with 2-methyl-amino pyridine.
Ramadan AM and El-Naggar MM.
Journal of Inorganic Biochemistry, 63(2), 143-153 (1996)
Hanna Rostkowska et al.
The journal of physical chemistry. A, 121(37), 6932-6941 (2017-08-30)
The UV-induced thione → thiol phototautomeric reaction has been studied for monomeric 3-thio-1,2,4-triazole (3-ST) isolated in low-temperature Ar and n-H
Giacomo Russo et al.
International journal of pharmaceutics, 560, 294-305 (2019-02-17)
The present work proposes a novel application of EPSA (not an acronym but found to be referred to by many as Exposed Polar Surface Area), a supercritical fluid chromatography (SFC) polarity readout for assisting in the prediction of the extent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service