Skip to Content
Merck
All Photos(2)

Key Documents

199524

Sigma-Aldrich

Chlorophenol Red

indicator grade

Synonym(s):

3′,3′-Dichlorophenolsulfonaphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12Cl2O5S
CAS Number:
Molecular Weight:
423.27
Beilstein:
354053
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

indicator grade

form

powder, crystals or chunks

color

dark brown

pH

4.8-6.7, yellow to violet

solubility

95% ethanol: 10 mg/mL

λmax

572 nm

ε (extinction coefficient)

≥12000 at 295-301 nm in 0.1 M NaOH
≥45000 at 573-579 nm in 0.1 M NaOH
≥5000 at 368-374 nm in 0.1 M NaOH

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1ccc(cc1Cl)C2(OS(=O)(=O)c3ccccc23)c4ccc(O)c(Cl)c4

InChI

1S/C19H12Cl2O5S/c20-14-9-11(5-7-16(14)22)19(12-6-8-17(23)15(21)10-12)13-3-1-2-4-18(13)27(24,25)26-19/h1-10,22-23H

InChI key

WWAABJGNHFGXSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Chlorophenol red is a pH indicator. It is used as an optical transducer of acetyl cholinesterase inhibition by the analytes. Chlorophenol red is also used to selectively determine chlorine dioxide in drinking water. It has been used in the novel assay to detect transformed protoplast-derived Zea mays colonies on Phosphinothricin (PPT).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fiber optic monitoring of carbamate pesticides using porous glass with covalently bound chlorophenol red.
Xavier MP
Biosensors And Bioelectronics, 14, 895-905 (2000)
Riccardo Martini et al.
Chembiochem : a European journal of chemical biology, 18(4), 374-377 (2016-12-20)
In recent years many advances have been made in the fight against HIV-1 infection. However, the lack of a vaccine, together with the increasing resistance to the highly active anti-retroviral therapy (HAART), make HIV-1 infection still a serious global emergency.
Federico Schaumburg et al.
Analytical chemistry, 91(15), 9623-9630 (2019-07-10)
Detecting bacteria is important in the fields of human health, environmental monitoring, and food safety. Foodborne pathogens alone are estimated to cause 420 000 deaths annually, with low-income regions affected most. Despite improvements in bacterial detection, fast, disposable, low-cost, sensitive, and
Julaluk Noiphung et al.
Theranostics, 8(14), 3797-3807 (2018-08-08)
Rationale: Saliva as a sample matrix is rapidly gaining interest for disease diagnosis and point-of-care assays because it is easy to collect (non-invasive) and contains many health-related biomarkers. However, saliva poses particular problems relative to more common urine and blood
Selection of transformed protoplast-derived Zea mays colonies with phosphinothricin and a novel assay using the pH indicator chlorophenol red
Kramer C
Planta, 190, 454-458 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service