Skip to Content
Merck
All Photos(2)

Documents

197017

Sigma-Aldrich

4-Chloro-3,5-dinitrobenzotrifluoride

98%

Synonym(s):

CDNT, DNT-Cl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(NO2)2CF3
CAS Number:
Molecular Weight:
270.55
Beilstein:
1220937
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

50-55 °C (lit.)

solubility

water: insoluble

SMILES string

[O-][N+](=O)c1cc(cc(c1Cl)[N+]([O-])=O)C(F)(F)F

InChI

1S/C7H2ClF3N2O4/c8-6-4(12(14)15)1-3(7(9,10)11)2-5(6)13(16)17/h1-2H

InChI key

HFHAVERNVFNSHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chloro-3,5-dinitrobenzotrifluoride was used:
  • as derivatization reagent in determination of amikacin in water samples and amino acids by HPLC
  • in the synthesis of N-(2,6-dinitro-4-trifluoromethylphenyl)aniline derivatives
  • in the synthesis of 4,6-dinitro-2,8-bis(trifluoromethyl)phenothiazine and isopropyl-2,6-dinitro-4-trifluoromethylphenyl sulfide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biren A Patel et al.
Journal of human evolution, 108, 176-198 (2017-06-18)
Songhor is an early Miocene fossil locality in Kenya known for its diverse primate assemblage that includes catarrhine species belonging to the genera Kalepithecus, Limnopithecus, Dendropithecus, Rangwapithecus, and Proconsul. Expeditions to Songhor since the 1930s have recovered unassociated catarrhine postcranial
S W Kamau et al.
BMC pharmacology, 1, 1-1 (2001-04-12)
Leishmaniasis is a major parasitic disease in the tropical regions. However, Leishmania infantum has recently emerged as a very important cause of opportunistic infections for individuals positive for human immunodeficiency virus (HIV). However, there is a lack of in vitro
Tianyu Shi et al.
Analytica chimica acta, 654(2), 154-161 (2009-10-27)
This work presents an high-performance liquid chromatography method for the determination of amino acids after precolumn derivatization with 4-chloro-3,5-dinitrobenzotrifluoride (CNBF) which can readily react with both primary and secondary amines. The precolumn derivatization conditions, including the CNBF concentration, reaction pH
Jawad M Khalifeh et al.
Neurosurgery, 84(2), 362-377 (2018-10-30)
Restoration of shoulder function is an important treatment goal in upper brachial plexus injury (UBPI). Combined dual motor nerve transfer (CDNT) of spinal accessory to suprascapular and radial to axillary nerves demonstrates good functional recovery with minimal risk of perioperative
Barbara Kappes et al.
Future microbiology, 2(4), 409-423 (2007-08-09)
The spread of parasitic resistance has necessitated the development of new drugs and drug targets for the treatment of malaria. Microtubules, which have gained outstanding importance as target molecules for the development of anticancer drugs, are likely to be potent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service