Recommended Products
form
powder
loss
≤15% loss on drying
Application
Lycopodium has been used as a hydrophobic coating on liquid marbles to make them non-adhering. It can be used as a pore-forming agent for the synthesis of porous hydroxyapatite and alumina ceramics.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mycopathologia, 185(4), 675-684 (2020-06-06)
Candida tropicalis is a non-albicans Candida specie that causes candidosis in several countries, including Brazil. However, little is known about the mechanisms of drug resistance in C. tropicalis infections. In this study, we used clinical isolates of C. tropicalis susceptible
Porous alumina ceramics produced with lycopodium spores as pore-forming agents.
J. Mater. Sci., 42(20), 8760-8764 (2007)
[Experience with the conditioned-reflex treatment of alcoholism with small doses of Lycopodium selago].
Vrachebnoe delo, (12)(12), 71-72 (1985-12-01)
Elasticity and failure of liquid marbles: influence of particle coating and marble volume.
Soft Matter, 13(47), 8903-8909 (2017)
Molecular plant, 13(11), 1644-1653 (2020-08-19)
The outer wall of pollen and spores, namely the exine, is composed of sporopollenin, which is highly resistant to chemical reagents and enzymes. In this study, we demonstrated that phenylpropanoid pathway derivatives are essential components of sporopollenin in seed plants.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service