Skip to Content
Merck
All Photos(2)

Key Documents

190993

Sigma-Aldrich

Poly(4-methyl-1-pentene)

medium molecular weight

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH[CH2CH(CH3)2])n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

melt index

26 g/10 min (260°C/5kg)

impact strength

3 kg.cm/cm2 (Izod, ASTM D 256)

dielectric constant

2.12, 100 MHz (ASTM D 150)

hardness

80 (Rockwell R, ASTM D 758)

refractive index

n20/D 1.463

mp

235 °C

transition temp

softening point 178 °C (Vicat, ASTM D 1525)

density

0.835 g/mL at 25 °C

SMILES string

CC(C)CC=C

InChI

1S/C6H12/c1-4-5-6(2)3/h4,6H,1,5H2,2-3H3

InChI key

WSSSPWUEQFSQQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coating for paper food containers for microwave and conventional ovens. Release coatings for food and synthetic leather. Molded into medical labware. Film for decorative laminates and printed circuit boards.

Features and Benefits

Very low specific gravity, high transparency, resistant to oxidation and prone to stress cracking. Good electrical insulator.

Physical form

Crystalline, isotactic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D M Wang et al.
Journal of controlled release : official journal of the Controlled Release Society, 50(1-3), 187-195 (1998-08-01)
In this study, asymmetric poly(4-methyl-1-pentene) (TPX) membranes, fabricated by the dry/wet inversion method, were applied to transdermal delivery of nitroglycerin (NTG), a drug for treating angina pectoris. The flux of NTG through the TPX membrane was measured in vitro by
Sang-Chul Shin et al.
International journal of pharmaceutics, 234(1-2), 67-73 (2002-02-13)
The pharmacokinetics and bioavailability of triprolidine, antihistamines, were studied to determine the feasibility of enhanced transdemal delivery of triprolidine from the poly(4-methyl-1-pentene) (TPX) matrix system containing polyoxyethylene-2-oleyl ether in rabbits. The triprolidine-TPX matrix (50 mg/kg) was applied to abdominal skin
LaNetra M Clayton et al.
Journal of nanoscience and nanotechnology, 6(8), 2520-2524 (2006-10-14)
Poly(4-methyl-1-pentene), PMP, a high melting polymer composed of hydrogen and carbon, has the potential to become an alternative to polyethylene (PE) as shielding material against Galactic Cosmic Radiation (GCR). PMP exhibits higher thermal stability than PE and is transparent in
Ayano Chiba et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 85(2 Pt 1), 021807-021807 (2012-04-03)
High-pressure in situ x-ray diffraction and specific-volume measurements on isotactic poly(4-methyl-1-pentene) melt have uncovered abrupt changes in the pressure dependence of microscopic structure as well as that of macroscopic density. The first sharp diffraction peak of the polymer melt, which
Sang-Chul Shin et al.
International journal of pharmaceutics, 235(1-2), 141-147 (2002-03-07)
Triprolidine-containing matrix was fabricated with poly(4-methyl-1-pentene) (TPX) polymer to control the release of the drug. Effect of penetration enhancer and stripping of skin on the permeation of triprolidine through the excised mouse skin was studied. Penetrating enhancers showed the increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service