Skip to Content
Merck
All Photos(2)

Key Documents

180823

Sigma-Aldrich

5-Methyl-5-phenylhydantoin

99%

Synonym(s):

5-Methyl-5-phenylimidazolidine-2,4-dione, NSC 14839

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O2
CAS Number:
Molecular Weight:
190.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

199-201 °C (lit.)

functional group

phenyl

separation technique

chiral

SMILES string

CC1(NC(=O)NC1=O)c2ccccc2

InChI

1S/C10H10N2O2/c1-10(7-5-3-2-4-6-7)8(13)11-9(14)12-10/h2-6H,1H3,(H2,11,12,13,14)

InChI key

JNGWGQUYLVSFND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
Beta-amino alcohols as inhibitors of anti-tubercular target N-acetyltransferase
Chlorohydantoins
Specific MMP inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel C Whitehead et al.
Tetrahedron letters, 50(6), 656-658 (2010-02-17)
A simple and efficient methodology for the preparation of N-chlorinated hydantoins is presented. These versatile chlorenium sources were isolated in high yield after a simple recrystallization. Among the ten examples are the first chiral N-chlorohydantoins.
Pirkle WH and Gan KZ.
Journal of Chromatography A, 790(1), 65-71 (1997)
Elizabeth Fullam et al.
Bioorganic & medicinal chemistry letters, 21(4), 1185-1190 (2011-01-22)
The synthesis and inhibitory potencies of a novel series of β-amino alcohols, based on the hit-compound 3-[3'-(4''-cyclopent-2'''-en-1'''-ylphenoxy)-2'-hydroxypropyl]-5,5 dimethylimidazolidine-2,4-dione as specific inhibitors of mycobacterial N-acetyltransferase (NAT) enzymes are reported. Effects of synthesised compounds on growth of Mycobacterium tuberculosis have been determined.
B Chankvetadz et al.
Journal of pharmaceutical and biomedical analysis, 15(9-10), 1577-1584 (1997-06-01)
The techniques for a dynamic and permanent reversal of the electroosmotic flow (EOF) were used for the reversal of the enantiomer migration order (EMO) of neutral and cationic analytes in chiral capillary electrophoresis (CE). Native beta-Cd and an anionic CD
Adriana Bakalova et al.
European journal of medicinal chemistry, 38(6), 627-632 (2003-07-02)
The interaction of cis-dichlorodiaminplatinum(II) (cis-DDP) with 2,4-imidazolidenedione-5-methyl-5-phenyl was studied. The method of preparation of the new Pt(II) complex consisted in precipitation of chloride ions from cis-DDP via a diaqua complex and reaction with the ligand in water-organic media. On the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service