Skip to Content
Merck
All Photos(1)

Key Documents

176737

Sigma-Aldrich

Nitric acid-d solution

65 wt. % in D2O, 99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DNO3
CAS Number:
Molecular Weight:
64.02
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

99 atom % D

form

solution

concentration

65 wt. % in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.3959

mass shift

M+1

SMILES string

[2H]O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)/i/hD

InChI key

GRYLNZFGIOXLOG-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service