Skip to Content
Merck
All Photos(3)

Key Documents

175560

Sigma-Aldrich

Triethoxyvinylsilane

97%

Synonym(s):

(Triethoxysilyl)ethylene, Vinyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHSi(OC2H5)3
CAS Number:
Molecular Weight:
190.31
Beilstein:
1767229
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

160-161 °C (lit.)
62-63 °C/20 mmHg (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)C=C

InChI

1S/C8H18O3Si/c1-5-9-12(8-4,10-6-2)11-7-3/h8H,4-7H2,1-3H3

InChI key

FWDBOZPQNFPOLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethoxyvinylsilane(TEVS) is a silane coupling agent and an adhesion promoter which can be used for cross-linking and for providing an insulating layer with good mechanical and thermal properties. [1][2]

Application

TEVS can be used in the surface modification of silica nanoparticles (SiNPs) which can be reinforced on linear-low density polyethylene (LLDP) for food packaging applications.[3] It can be encapsulated with different pigments like protoporphrin XI[4] and methylene blue[5] to artificially sensitize the tissues for photodynamic therapy.
Triethoxyvinylsilane (TEVS) was used to modify nano-silica particles[6]and poly (vinyl acetate). [7] TEVS may undergo emulsion copolymerization with styrene (St), butyl acrylate (BA), and methacrylic acid (MAA).[8] The formation of random copolymers of TEVS with styrene (St) has been reported.[9] TEVS may undergo graft polymerization with styrene and methyl methacrylate over different silicates .[10] [11]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pre-treatment of flax fibers for use in rotationally molded biocomposites
Wang, B., Panigrahi, S., Tabil, L., & Crerar, W.
Journal of Reinforced Plastics and Composites, 26(5), 447-463 (2007)
Effect of nano-silica loading on barrier and mechanical properties of food packaging based LLDPE film.
Thanakkasaranee S, et al.
Advanced Materials Research, 488, 919-922 (2012)
Process optimization for improved adhesion in the grafting of triethoxyvinylsilane on VLDPE via reactive extrusion.
Schaible S, et al.
European Polymer Journal, 107, 9-14 (2018)
Effect of Photon Radiations in Semi-Rigid Artificial Tissue Sensitized by Protoporphyrin IX Encapsulated with Silica Nanoparticles.
IOP Conference Series: Materials Science and Engineering, 305(1), 012014-012014 (2018)
Preparation and Properties of Triethoxyvinylsilane-Modified Styrene-Butyl Acrylate Emulsion Copolymers
Turkish Journal of Chemistry, 31(3), 257-257 (2007)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service