Skip to Content
Merck
All Photos(1)

Key Documents

170216

Sigma-Aldrich

Methylglyoxal 1,1-dimethyl acetal

≥97%

Synonym(s):

1,1-Dimethoxyacetone, Pyruvaldehyde-1-dimethyl acetal, Pyruvic aldehyde dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(OCH3)2
CAS Number:
Molecular Weight:
118.13
Beilstein:
1560557
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

143-147 °C (lit.)

density

0.976 g/mL at 25 °C (lit.)

functional group

acetal
ether
ketone

SMILES string

COC(OC)C(C)=O

InChI

1S/C5H10O3/c1-4(6)5(7-2)8-3/h5H,1-3H3

InChI key

ULVSHNOGEVXRDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methylglyoxal 1,1-dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase in vivo.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetlana I Dorovskikh et al.
Biomedicines, 9(8) (2021-08-28)
This work is aimed at developing the modification of the surface of medical implants with film materials based on noble metals in order to improve their biological characteristics. Gas-phase transportation methods were proposed to obtain such materials. To determine the
Ajit Ghosh et al.
Scientific reports, 6, 18358-18358 (2016-01-07)
Glyoxalase pathway, comprising glyoxalase I (GLY I) and glyoxalase II (GLY II) enzymes, is the major pathway for detoxification of methylglyoxal (MG) into D-lactate involving reduced glutathione (GSH). However, in bacteria, glyoxalase III (GLY III) with DJ-1/PfpI domain(s) can do
K Ispolnov et al.
Journal of applied microbiology, 104(4), 1092-1102 (2008-01-16)
The purpose of this study was to investigate the behaviour of Saccharomyces cerevisiae in response to extracellular methylglyoxal. Cell survival to methylglyoxal and the importance of phosphates was investigated. The role of methylglyoxal detoxification systems and methylglyoxal-derived protein glycation were
Ricardo A Gomes et al.
The FEBS journal, 272(17), 4521-4531 (2005-09-01)
Methylglyoxal is the most important intracellular glycation agent, formed nonenzymatically from triose phosphates during glycolysis in eukaryotic cells. Methylglyoxal-derived advanced glycation end-products are involved in neurodegenerative disorders (Alzheimer's, Parkinson's and familial amyloidotic polyneurophathy) and in the clinical complications of diabetes.
Lokman Liv et al.
Analytical and bioanalytical chemistry, 414(3), 1313-1322 (2021-11-07)
Among the deadliest pandemics in history, coronavirus disease 2019 (COVID-19) has wreaked havoc on human lives, economies and public health systems worldwide. To temper its effects, diagnostic methods that are simple, rapid, inexpensive, accurate, selective and sensitive continue to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service